accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

O

CAS RN 667-91-4

IUPAC Name: 10-(diphenylmethylidene)-9,10-dihydroanthracen-9-one
Synonyms: 10-(diphenylmethylidene)anthracen-9-one
Molecular Weight (g/mol): 358.44
Molecular Formula: C27H18O
InChi Key: SPOJSTUMWGMCFP-UHFFFAOYSA-N
SMILES: O=C1C2=CC=CC=C2C(=C(C2=CC=CC=C2)C2=CC=CC=C2)C2=CC=CC=C12
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results