accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CCH3H3CCH3

CAS RN 6683-46-1

IUPAC Name: 1,1,4,4-tetramethyl-1,2,3,4-tetrahydronaphthalene
Synonyms: 1,1,4,4-tetramethyl-2,3-dihydronaphthalene
Molecular Weight (g/mol): 188.31
Molecular Formula: C14H20
InChi Key: CCQKWSZYTOCEIB-UHFFFAOYSA-N
SMILES: CC1(C)CCC(C)(C)C2=CC=CC=C12
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (1)