accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BSHOOH

CAS RN 668983-97-9

IUPAC Name: {8-thiatricyclo[7.4.0.0²,⁷]trideca-1(9),2(7),3,5,10,12-hexaen-4-yl}boronic acid
Synonyms: 8-thiatricyclo[7.4.0.0²,⁷]trideca-1(9),2(7),3,5,10,12-hexaen-4-ylboronic acid
Molecular Weight (g/mol): 228.07
Molecular Formula: C12H9BO2S
InChi Key: CSLSCVHILGCSTE-UHFFFAOYSA-N
SMILES: OB(O)C1=CC2=C(SC3=C2C=CC=C3)C=C1
Molecular Weight (g/mol) 
  • (3)
  • (6)
Quantity 
  • (2)
  • (1)
  • (3)
  • (2)
  • (1)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (6)
  • (2)
  • (1)
  • (3)
  • (2)
  • (1)
  • (3)
  • (3)
  • (3)