accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 66905-23-5

IUPAC Name: calcium bis(2,3,4,5,6-pentahydroxyhexanoate) hydrate
Synonyms: calcium bis(2,3,4,5,6-pentahydroxyhexanoate) hydrate
Molecular Weight (g/mol): 448.39
Molecular Formula: C12H24CaO15
InChi Key: XLNFVCRGJZBQGX-UHFFFAOYNA-L
SMILES: O.[Ca++].OCC(O)C(O)C(O)C(O)C([O-])=O.OCC(O)C(O)C(O)C(O)C([O-])=O
Molecular Weight (g/mol) 
  • (1)
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results