accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 669050-70-8

IUPAC Name: 1H-indazole-4-carbaldehyde
Synonyms: 1H-indazole-4-carbaldehyde
Molecular Weight (g/mol): 146.15
Molecular Formula: C8H6N2O
InChi Key: FPJXNCKSPFGQGC-UHFFFAOYSA-N
SMILES: O=CC1=C2C=NNC2=CC=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results