accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 66943-05-3

IUPAC Name: 1,4,7,10-tetraoxa-13-azacyclopentadecan-13-ium
Synonyms: 1,4,7,10-tetraoxa-13-azacyclopentadecan-13-ium
Molecular Weight (g/mol): 220.29
Molecular Formula: C10H22NO4
InChi Key: BJUOQSZSDIHZNP-UHFFFAOYSA-O
SMILES: C1COCCOCCOCCOCC[NH2+]1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results