CAS RN 67-03-8
Thermo Scientific Chemicals Thiamine hydrochloride, 98.5-101.5%
CAS: 67-03-8 Molecular Formula: C12H17ClN4OS·HCl Molecular Weight (g/mol): 337.26 MDL Number: MFCD00012780 InChI Key: DPJRMOMPQZCRJU-UHFFFAOYSA-M Synonym: thiamine hydrochloride,vitamin b1 hydrochloride,thiamine hcl,aneurine hydrochloride,berin,thiamine chloride hydrochloride,trophite,thiamin chloride,thiamine dichloride,thiaminium chloride PubChem CID: 6202 ChEBI: CHEBI:49105 IUPAC Name: 2-[3-[(4-amino-2-methylpyrimidin-5-yl)methyl]-4-methyl-1,3-thiazol-3-ium-5-yl]ethanol;chloride;hydrochloride SMILES: CC1=C(SC=[N+]1CC2=CN=C(N=C2N)C)CCO.Cl.[Cl-]
Thiamine hydrochloride, 99% (dry wt.), may cont. up to 5% water
CAS: 67-03-8 Molecular Formula: C12H18Cl2N4OS Molecular Weight (g/mol): 337.263 MDL Number: MFCD00012780 InChI Key: DPJRMOMPQZCRJU-UHFFFAOYSA-M Synonym: thiamine hydrochloride,vitamin b1 hydrochloride,thiamine hcl,aneurine hydrochloride,berin,thiamine chloride hydrochloride,trophite,thiamin chloride,thiamine dichloride,thiaminium chloride PubChem CID: 6202 ChEBI: CHEBI:49105 IUPAC Name: 2-[3-[(4-amino-2-methylpyrimidin-5-yl)methyl]-4-methyl-1,3-thiazol-3-ium-5-yl]ethanol;chloride;hydrochloride SMILES: CC1=C(SC=[N+]1CC2=CN=C(N=C2N)C)CCO.Cl.[Cl-]
Thiamine Hydrochloride, Powder, MP Biomedicals
CAS: 67-03-8 Molecular Formula: C12H17ClN4OS·HCl Molecular Weight (g/mol): 337.263 MDL Number: MFCD00012780 InChI Key: UIERGBJEBXXIGO-UHFFFAOYSA-N Synonym: Aneurine hydrochloride,Vitamin B1 hydrochloride,Vitamin B1 HCl PubChem CID: 10762 SMILES: [O-][N+]([O-])=O.CC1=C(CCO)[S+]=CN1CC1=CN=C(C)N=C1N
Thiamine hydrochloride 10 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
Thiamine Hydrochloride (Vitamin B1), Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Thiamine Hydrochloride (Vitamin B1) 1.0 mg/ml in Methanol (as free base), LoGiCal
LoGiCal delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
Thiamine Hydrochloride, TRC
CAS: 67-03-8 Molecular Formula: C12H17N4OS.Cl.HCl Molecular Weight (g/mol): 337.27 Synonym: Thiamine Hydrochloride,Vitamin B1,Thiamine monohydrochloride,Thiazolium, 3-[(4-amino-2-methyl-5-pyrimidinyl)methyl]-5-(2-hydroxyethyl)-4-methyl-, chloride, hydrochloride (1:1:1),Thiamine, monohydrochloride (8CI),Thiazolium, 3-[(4-amino-2-methyl-5-pyrimidinyl)methyl]-5-(2-hydroxyethyl)-4-methyl-, chloride, monohydrochloride (9CI),3-[(4-Amino-2-methyl-5-pyrimidinyl)methyl]-5-(2-hydroxyethyl)-4-methylthiazolium chloride monohydrochloride,3-(4-Amino-2-methylpyrimidyl-5-methyl)-4-methyl-5,β-hydroxyethylthiazolium chloride hydrochloride,Aneurine hydrochloride,Apate drops,Beatine,Bedome,Begiolan,Benerva,Bequin,Berin,Betabion,Betalin S,Betaxin,Bethiazine,Beuion,Bevitine,Bewon,Biamine,Biuno,Bivatin,Bivita,Clotiamina,Eskapen,Eskaphen,Lixa-Beta,Metabolin,Slowten,THD,Thiadoxine,Thiamin chloride,Thiamin dichloride,Thiamin hydrochloride,Thiaminal,Thiamine chloride,Thiamine chloride hydrochloride,Thiamine dichloride,Thiamine hydrochloride,Thiaminium chloride,Thiaminium chloride hydrochloride,Thiamol,Thiavit,Tiamidon,Tiaminal,Trophite,Vetalin S,Vinothiam,Vitamin B1 hydrochloride,Vitaneuron IUPAC Name: 2-[3-[(4-amino-2-methylpyrimidin-5-yl)methyl]-4-methyl-1,3-thiazol-3-ium-5-yl]ethanol;chloride;hydrochloride SMILES: Cl.[Cl-].Cc1ncc(C[n+]2csc(CCO)c2C)c(N)n1
Thiamine Hydrochloride (Vitamin B1), LoGiCal
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
Thiamine hydrochloride, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
Thiamine Hydrochloride, U.S.P. - F.C.C., J.T. Baker™
CAS: 67-03-8 Molecular Formula: C12H18Cl2N4OS Molecular Weight (g/mol): 337.263 InChI Key: DPJRMOMPQZCRJU-UHFFFAOYSA-M Synonym: thiamine hydrochloride,vitamin b1 hydrochloride,thiamine hcl,aneurine hydrochloride,berin,thiamine chloride hydrochloride,trophite,thiamin chloride,thiamine dichloride,thiaminium chloride PubChem CID: 6202 ChEBI: CHEBI:49105 IUPAC Name: 2-[3-[(4-amino-2-methylpyrimidin-5-yl)methyl]-4-methyl-1,3-thiazol-3-ium-5-yl]ethanol;chloride;hydrochloride SMILES: CC1=C(SC=[N+]1CC2=CN=C(N=C2N)C)CCO.Cl.[Cl-]