CAS RN 67-20-9
Nitrofurantoin, 98%
CAS: 67-20-9 Molecular Formula: C8H6N4O5 Molecular Weight (g/mol): 238.16 MDL Number: MFCD00003224 InChI Key: NXFQHRVNIOXGAQ-YCRREMRBSA-N Synonym: nitrofurantoin,macrodantin,5-nitrofurantoin,furadantin,furadonine,nitrofurantoine,furadantine,furadantoin,furadoine,furadontin PubChem CID: 6604200 ChEBI: CHEBI:71415 SMILES: [O-][N+](=O)C1=CC=C(O1)\C=N\N1CC(=O)NC1=O
Nitrofurantoin, 98%
CAS: 67-20-9 Molecular Formula: C8H6N4O5 Molecular Weight (g/mol): 238.16 MDL Number: MFCD00003224 InChI Key: NXFQHRVNIOXGAQ-YCRREMRBSA-N Synonym: nitrofurantoin,macrodantin,5-nitrofurantoin,furadantin,furadonine,nitrofurantoine,furadantine,furadantoin,furadoine,furadontin PubChem CID: 6604200 ChEBI: CHEBI:71415 IUPAC Name: 1-[(E)-[(5-nitrofuran-2-yl)methylidene]amino]imidazolidine-2,4-dione SMILES: [O-][N+](=O)C1=CC=C(O1)\C=N\N1CC(=O)NC1=O
Nitrofurantoin, TRC
CAS: 67-20-9 Molecular Formula: C8 H6 N4 O5 Molecular Weight (g/mol): 238.16 Synonym: Nitrofurantoin,1-[[(5-Nitro-2-furanyl)methylene]amino]-2,4-imidazolidinedione,1-(5-Nitro-2-furfurylidenamino)hydantoin,1-(5-Nitro-2-furfurylideneamino)hydantoin,1-[(5-Nitrofurfurylidene)amino]hydantoin,5-Nitrofurantoin,Berkfurin,Chemiofuran,Cyantin,Cystit,FuaMed,Fur-ren,Furachel,Furadantin,Furadantine MC,Furadantoin,Furadoin,Furadoine,Furadonin,Furadonine,Furadontin,Furalan,Furantoin,Furantoina,Furatoin,Furina,Furobactina,Furophen T-Caps,Ituran,Macrobid,Macrodantin,N-(5-Nitro-2-furfurylidene)-1-aminohydantoin,N-(5-Nitro-2-furfurylidine)-1-aminohydantoin,N-(5-Nitrofurfurylidene)-1-aminohydantoin,NSC 2107,NSC 44150,Nifurantin,Nitoin,Novofuran,Orafuran,Parfuran,Trantoin,Urantoin,Urizept,Uro-Tablinen,Urodin,Urofurin,Urolong,Welfurin,Zoofurin IUPAC Name: 1-[(E)-(5-nitrofuran-2-yl)methylideneamino]imidazolidine-2,4-dione SMILES: [O-][N+](=O)c1oc(\C=N\N2CC(=O)NC2=O)cc1
Nitrofurantoin, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
Nitrofurantoin, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Nitrofurantoin, LoGiCal
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
Melford Nitrofurantoin
Nitrofurantoin works by inhibiting bacterial enzymes involved in the metabolism of certain compounds, leading to the disruption of bacterial DNA, RNA, and protein synthesis.
Nitrofurantoin Yellow crystals. MP Biomedicals
CAS: 67-20-9 Molecular Formula: C8H6N4O5 Molecular Weight (g/mol): 238.16 MDL Number: MFCD00003224 InChI Key: NXFQHRVNIOXGAQ-YCRREMRBSA-N Synonym: nitrofurantoin,macrodantin,5-nitrofurantoin,furadantin,furadonine,nitrofurantoine,furadantine,furadantoin,furadoine,furadontin PubChem CID: 6604200 ChEBI: CHEBI:71415 SMILES: [O-][N+](=O)C1=CC=C(O1)\C=N\N1CC(=O)NC1=O