accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 67-36-7

IUPAC Name: 4-phenoxybenzaldehyde
Synonyms: benzaldehyde, p-phenoxy-
Molecular Weight (g/mol): 198.22
Molecular Formula: C13H10O2
InChi Key: QWLHJVDRPZNVBS-UHFFFAOYSA-N
SMILES: O=CC1=CC=C(OC2=CC=CC=C2)C=C1
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)
  • (4)
  • (3)
Percent Purity 
  • (5)
  • (5)
Boiling Point 
  • (3)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (1)
  • (3)
  • (3)
  • (5)
  • (2)
  • (2)
  • (1)
  • (1)
  • (4)
  • (3)
  • (5)
  • (5)
  • (3)
  • (2)
  • (2)