accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 67-43-6

IUPAC Name: 2-[bis({2-[bis(carboxymethyl)amino]ethyl})amino]acetic acid
Synonyms: DTPA pentacine pentetic acid (product) calcium-DTPA pentetic acid (substance) chel dtpa ca-dtpa calcium dtpa Sn-DTPA dissolvine D
Molecular Weight (g/mol): 393.35
Molecular Formula: C14H23N3O10
InChi Key: QPCDCPDFJACHGM-UHFFFAOYSA-N
SMILES: OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CCN(CC(O)=O)CC(O)=O
Molecular Weight (g/mol) 
  • (3)
  • (5)
  • (2)
Percent Purity 
  • (3)
  • (2)
  • (1)
  • (1)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (3)
  • (5)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (2)