accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ONOONNOO(E)

CAS RN 67-45-8

IUPAC Name: 3-[(E)-[(5-nitrofuran-2-yl)methylidene]amino]-1,3-oxazolidin-2-one
Synonyms: furazolidone ortazol furox viofuragyn neftin furaxone furall furazolidon furazolidona 呋喃唑酮
Molecular Weight (g/mol): 225.16
Molecular Formula: C8H7N3O5
InChi Key: PLHJDBGFXBMTGZ-WEVVVXLNSA-N
SMILES: [O-][N+](=O)C1=CC=C(O1)\C=N\N1CCOC1=O
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)