accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ClCH3NCH3CH3HO

CAS RN 67-48-1

IUPAC Name: (2-hydroxyethyl)trimethylazanium chloride
Synonyms: choline chloride biocolina biocoline hepacholine cholinium chloride cholini chloridum chlorure de choline bilineurin chloride cloruro de colina choline hydrochloride
Molecular Weight (g/mol): 139.62
Molecular Formula: C5H14ClNO
InChi Key: SGMZJAMFUVOLNK-UHFFFAOYSA-M
SMILES: [Cl-].C[N+](C)(C)CCO
Quantity 
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (4)
  • (1)
Percent Purity 
  • (1)
  • (4)
  • (2)
  • (2)
  • (3)
  • (4)
Physical Form 
  • (4)
  • (6)
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (7)
  • (2)
  • (4)
  • (3)
  • (3)
  • (5)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (4)
  • (1)
  • (1)
  • (4)
  • (2)
  • (2)
  • (3)
  • (4)
  • (4)
  • (6)
  • (2)
  • (3)
  • (1)
  • (1)