accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OONHOHN

CAS RN 67-52-7

IUPAC Name: 1,3-diazinane-2,4,6-trione
Synonyms: barbituric acid malonylharnstoff barbitursaeure
Molecular Weight (g/mol): 128.09
Molecular Formula: C4H4N2O3
InChi Key: HNYOPLTXPVRDBG-UHFFFAOYSA-N
SMILES: O=C1CC(=O)NC(=O)N1
Quantity 
  • (2)
  • (1)
  • (2)