CAS RN 67-63-0
Isopropanol, Certified AR for Analysis, Fisher Chemical™
CAS: 67-63-0 Molecular Formula: C3H8O Molecular Weight (g/mol): 60.096 MDL Number: 11674 InChI Key: KFZMGEQAYNKOFK-UHFFFAOYSA-N Synonym: isopropanol,2-propanol,isopropyl alcohol,2-hydroxypropane,alkolave,avantine,hartosol,dimethylcarbinol,sec-propyl alcohol,petrohol PubChem CID: 3776 ChEBI: CHEBI:17824 IUPAC Name: propan-2-ol SMILES: CC(C)O
Isopropanol, 99.5+%, pure
CAS: 67-63-0 Molecular Formula: C3H8O Molecular Weight (g/mol): 60.096 MDL Number: MFCD00011674 InChI Key: KFZMGEQAYNKOFK-UHFFFAOYSA-N Synonym: isopropanol,2-propanol,isopropyl alcohol,2-hydroxypropane,alkolave,avantine,hartosol,dimethylcarbinol,sec-propyl alcohol,petrohol PubChem CID: 3776 ChEBI: CHEBI:17824 IUPAC Name: propan-2-ol SMILES: CC(C)O
Isopropanol, Optima™LC/MS Grade, Fisher Chemical™
CAS: 67-63-0 Molecular Formula: C3H8O Molecular Weight (g/mol): 60.10 MDL Number: 11674 InChI Key: KFZMGEQAYNKOFK-UHFFFAOYSA-N Synonym: isopropanol,2-propanol,isopropyl alcohol,2-hydroxypropane,alkolave,avantine,hartosol,dimethylcarbinol,sec-propyl alcohol,petrohol PubChem CID: 3776 ChEBI: CHEBI:17824 IUPAC Name: propan-2-ol SMILES: CC(C)O
Isopropanol, Extra Pure, SLR, Fisher Chemical™
CAS: 67-63-0 Molecular Formula: C3H8O Molecular Weight (g/mol): 60.096 MDL Number: 11674 InChI Key: KFZMGEQAYNKOFK-UHFFFAOYSA-N Synonym: isopropanol,2-propanol,isopropyl alcohol,2-hydroxypropane,alkolave,avantine,hartosol,dimethylcarbinol,sec-propyl alcohol,petrohol PubChem CID: 3776 ChEBI: CHEBI:17824 IUPAC Name: propan-2-ol SMILES: CC(C)O
Isopropanol, for HPLC, Fisher Chemical™
CAS: 67-63-0 Molecular Formula: C3H8O Molecular Weight (g/mol): 60.10 MDL Number: 11674 InChI Key: KFZMGEQAYNKOFK-UHFFFAOYSA-N Synonym: isopropanol,2-propanol,isopropyl alcohol,2-hydroxypropane,alkolave,avantine,hartosol,dimethylcarbinol,sec-propyl alcohol,petrohol PubChem CID: 3776 ChEBI: CHEBI:17824 IUPAC Name: propan-2-ol SMILES: CC(C)O
2-Propanol, Molecular Biology Grade, Fisher BioReagents™
CAS: 67-63-0 Molecular Formula: C3H8O Molecular Weight (g/mol): 60.10 InChI Key: KFZMGEQAYNKOFK-UHFFFAOYSA-N Synonym: isopropanol,2-propanol,isopropyl alcohol,2-hydroxypropane,alkolave,avantine,hartosol,dimethylcarbinol,sec-propyl alcohol,petrohol PubChem CID: 3776 ChEBI: CHEBI:17824 IUPAC Name: propan-2-ol SMILES: CC(C)O
Isopropanol, 99.8%, Extra Dry, AcroSeal™
CAS: 67-63-0 Molecular Formula: C3H8O Molecular Weight (g/mol): 60.096 MDL Number: MFCD00011674 InChI Key: KFZMGEQAYNKOFK-UHFFFAOYSA-N Synonym: isopropanol,2-propanol,isopropyl alcohol,2-hydroxypropane,alkolave,avantine,hartosol,dimethylcarbinol,sec-propyl alcohol,petrohol PubChem CID: 3776 ChEBI: CHEBI:17824 IUPAC Name: propan-2-ol SMILES: CC(C)O
Isopropanol, 99.5+%, extra pure
CAS: 67-63-0 Molecular Formula: C3H8O Molecular Weight (g/mol): 60.096 MDL Number: MFCD00011674 InChI Key: KFZMGEQAYNKOFK-UHFFFAOYSA-N Synonym: isopropanol,2-propanol,isopropyl alcohol,2-hydroxypropane,alkolave,avantine,hartosol,dimethylcarbinol,sec-propyl alcohol,petrohol PubChem CID: 3776 ChEBI: CHEBI:17824 IUPAC Name: propan-2-ol SMILES: CC(C)O
Isopropanol, 99.6%, ACS reagent, meets the requirements of Reag.Ph.Eur.
CAS: 67-63-0 Molecular Formula: C3H8O Molecular Weight (g/mol): 60.096 MDL Number: MFCD00011674 InChI Key: KFZMGEQAYNKOFK-UHFFFAOYSA-N Synonym: isopropanol,2-propanol,isopropyl alcohol,2-hydroxypropane,alkolave,avantine,hartosol,dimethylcarbinol,sec-propyl alcohol,petrohol PubChem CID: 3776 ChEBI: CHEBI:17824 IUPAC Name: propan-2-ol SMILES: CC(C)O
Isopropanol, 99.5%, for HPLC
CAS: 67-63-0 Molecular Formula: C3H8O Molecular Weight (g/mol): 60.10 MDL Number: MFCD00011674 InChI Key: KFZMGEQAYNKOFK-UHFFFAOYSA-N Synonym: isopropanol,2-propanol,isopropyl alcohol,2-hydroxypropane,alkolave,avantine,hartosol,dimethylcarbinol,sec-propyl alcohol,petrohol PubChem CID: 3776 ChEBI: CHEBI:17824 IUPAC Name: propan-2-ol SMILES: CC(C)O
2-Propanol, GC Grade, 99.7+%
CAS: 67-63-0 Molecular Formula: C3H8O Molecular Weight (g/mol): 60.10 MDL Number: MFCD00011674 InChI Key: KFZMGEQAYNKOFK-UHFFFAOYSA-N Synonym: isopropanol,2-propanol,isopropyl alcohol,2-hydroxypropane,alkolave,avantine,hartosol,dimethylcarbinol,sec-propyl alcohol,petrohol PubChem CID: 3776 ChEBI: CHEBI:17824 IUPAC Name: propan-2-ol SMILES: CC(C)O
Isopropanol, 99.5%, for molecular biology, DNAse, RNAse and Protease free
CAS: 67-63-0 Molecular Formula: C3H8O Molecular Weight (g/mol): 60.096 MDL Number: MFCD00011674 InChI Key: KFZMGEQAYNKOFK-UHFFFAOYSA-N Synonym: isopropanol,2-propanol,isopropyl alcohol,2-hydroxypropane,alkolave,avantine,hartosol,dimethylcarbinol,sec-propyl alcohol,petrohol PubChem CID: 3776 ChEBI: CHEBI:17824 IUPAC Name: propan-2-ol SMILES: CC(C)O
Isopropanol, 99.5%, for analysis
CAS: 67-63-0 Molecular Formula: C3H8O Molecular Weight (g/mol): 60.096 MDL Number: MFCD00011674 InChI Key: KFZMGEQAYNKOFK-UHFFFAOYSA-N Synonym: isopropanol,2-propanol,isopropyl alcohol,2-hydroxypropane,alkolave,avantine,hartosol,dimethylcarbinol,sec-propyl alcohol,petrohol PubChem CID: 3776 ChEBI: CHEBI:17824 IUPAC Name: propan-2-ol SMILES: CC(C)O
Isopropanol, 99.5%, Extra Dry over Molecular Sieve, AcroSeal™
CAS: 67-63-0 Molecular Formula: C3H8O MDL Number: MFCD00011674 InChI Key: KFZMGEQAYNKOFK-UHFFFAOYSA-N Synonym: isopropanol,2-propanol,isopropyl alcohol,2-hydroxypropane,alkolave,avantine,hartosol,dimethylcarbinol,sec-propyl alcohol,petrohol PubChem CID: 3776 ChEBI: CHEBI:17824 IUPAC Name: propan-2-ol SMILES: CC(C)O