accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 670-54-2

NNNN

CAS RN 670-54-2

IUPAC Name: eth-1-ene-1,1,2,2-tetracarbonitrile
Synonyms: tetracyanoethylene ethenetetracarbonitrile
Molecular Weight (g/mol): 128.09
Molecular Formula: C6N4
InChi Key: NLDYACGHTUPAQU-UHFFFAOYSA-N
SMILES: N#CC(C#N)=C(C#N)C#N
Boiling Point 
  • (2)
  • (1)
  • (3)
Molecular Weight (g/mol) 
  • (4)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (4)
  • (2)
  • (2)
  • (1)
  • (3)
  • (4)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (4)