accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 670-80-4

IUPAC Name: 4-(cyclohex-1-en-1-yl)morpholine
Synonyms: 4-(cyclohex-1-en-1-yl)morpholine
Molecular Weight (g/mol): 167.25
Molecular Formula: C10H17NO
InChi Key: IIQFBBQJYPGOHJ-UHFFFAOYSA-N
SMILES: C1CCC(=CC1)N1CCOCC1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)