accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 670-98-4

CH3OSO

CAS RN 670-98-4

IUPAC Name: methyl benzenesulfinate
Synonyms: methyl benzenesulfinate
Molecular Weight (g/mol): 156.2
Molecular Formula: C7H8O2S
InChi Key: PSNSVDSRLUYDKF-UHFFFAOYNA-N
SMILES: COS(=O)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)