accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 67029-84-9

IUPAC Name: 6-chloro-7-methyl-4H-chromen-4-one
Synonyms: 6-chloro-7-methylchromen-4-one
Molecular Weight (g/mol): 194.61
Molecular Formula: C10H7ClO2
InChi Key: UQXYHMICNLSDMN-UHFFFAOYSA-N
SMILES: CC1=C(Cl)C=C2C(=O)C=COC2=C1
Molecular Weight (g/mol) 
  • (4)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (4)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)