accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 6704-31-0

IUPAC Name: oxetan-3-one
Synonyms: 3-oxetanone
Molecular Weight (g/mol): 72.06
Molecular Formula: C3H4O2
InChi Key: ROADCYAOHVSOLQ-UHFFFAOYSA-N
SMILES: O=C1COC1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results