CAS RN 6709-39-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Geraniolene, TRC
CAS: 6709-39-3 Molecular Formula: C9H16 Molecular Weight (g/mol): 124.22 Synonym: 2,6-Dimethylhepta-1,5-diene IUPAC Name: 2,6-dimethylhepta-1,5-diene SMILES: CC(=CCCC(=C)C)C