CAS RN 671-95-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Clofenamide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Clofenamide, TRC
CAS: 671-95-4 Molecular Formula: C6 H7 Cl N2 O4 S2 Molecular Weight (g/mol): 270.71 Synonym: 4-Chloro-1,3-benzenedisulfonamide,4-Chloro-m-benzenedisulfonamide,4-Chloro-m-benzenedisulfonamide,4-Chlorobenzene-1,3-disulfonamide,Aponiere,Aquedux,Clofenamidum,Diumide,Diuretic 2822,Eleklin,Frictan,Haflutan,Indigatin,Macashi,Monochlorphenamide,Salco,Saltron,Salzen,Soluran; IUPAC Name: 4-chlorobenzene-1,3-disulfonamide SMILES: NS(=O)(=O)c1ccc(Cl)c(c1)S(=O)(=O)N