accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 6714-68-7

ONHOO

CAS RN 6714-68-7

IUPAC Name: 1',2'-dihydrospiro[1,3-dioxolane-2,3'-indol]-2'-one
Synonyms: 1'H-spiro[1,3-dioxolane-2,3'-indol]-2'-one
Molecular Weight (g/mol): 191.19
Molecular Formula: C10H9NO3
InChi Key: PRMHWSVVQZVDGR-UHFFFAOYSA-N
SMILES: O=C1NC2=CC=CC=C2C11OCCO1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results