accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

SFFFFFHO

CAS RN 672-31-1

IUPAC Name: 3-(pentafluoro-λ⁶-sulfanyl)phenol
Synonyms: 3-(pentafluoro-λ⁶-sulfanyl)phenol
Molecular Weight (g/mol): 220.16
Molecular Formula: C6H5F5OS
InChi Key: KVOACEHJTNSNBD-UHFFFAOYSA-N
SMILES: OC1=CC(=CC=C1)S(F)(F)(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results