accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COHOOHNH2

CAS RN 672-86-6

IUPAC Name: 2-amino-3-(4-hydroxyphenyl)-2-methylpropanoic acid
Synonyms: AMPT
Molecular Weight (g/mol): 195.22
Molecular Formula: C10H13NO3
InChi Key: NHTGHBARYWONDQ-UHFFFAOYNA-N
SMILES: CC(N)(CC1=CC=C(O)C=C1)C(O)=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results