accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2NS

CAS RN 672324-88-8

IUPAC Name: 1-[3-(1,3-thiazol-2-yl)phenyl]methanamine
Synonyms: 1-[3-(1,3-thiazol-2-yl)phenyl]methanamine
Molecular Weight (g/mol): 190.26
Molecular Formula: C10H10N2S
InChi Key: UGCRMJJXRCJZHJ-UHFFFAOYSA-N
SMILES: NCC1=CC=CC(=C1)C1=NC=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results