accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

IN

CAS RN 6727-73-7

IUPAC Name: 4-iodobutanenitrile
Synonyms: 4-iodobutanenitrile
Molecular Weight (g/mol): 195
Molecular Formula: C4H6IN
InChi Key: CWINCVBWHUGBEB-UHFFFAOYSA-N
SMILES: ICCCC#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results