accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 673-40-5

BrNNFBFFF

CAS RN 673-40-5

IUPAC Name: boron(3+) 4-bromobenzene-1-diazonium tetrafluoride
Synonyms: boron(3+) 4-bromobenzenediazonium tetrafluoride
Molecular Weight (g/mol): 270.82
Molecular Formula: C6H4BBrF4N2
InChi Key: HSHRIOFZCXDGPR-UHFFFAOYSA-J
SMILES: [B+3].[F-].[F-].[F-].[F-].BrC1=CC=C(C=C1)[N+]#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results