CAS RN 6730-83-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
ent-Kaurenoic Acid, TRC
CAS: 6730-83-2 Molecular Formula: C20H30O2 Molecular Weight (g/mol): 302.45 Synonym: (4α)-Kaur-16-en-18-oic Acid,(-)-Kaur-16-en-19-oic Acid,(-)-Kauren-19-oic Acid,Cunabic Acid,Kaurenic Acid,Kaurenoic Acid,NSC 339145,ent-Kaur-16(17)-en-19-oic Acid,ent-Kaur-16-en-19-oic Acid,(4α)-Kaur-16-en-18-oic Acid SMILES: C=C1C[C@@]23CC[C@]4([H])[C@](C)(C(O)=O)CCC[C@@]4(C)[C@]2([H])CC[C@@H]1C3