CAS RN 67330-25-0
IUPAC Name:
butyl 2-{[3-(trifluoromethyl)phenyl]amino}benzoate
Synonyms:
combec flufenamic acid butyl ester ufenamatum butyl flufenamate ufenamato ufenamate
Molecular Weight (g/mol):
337.34
Molecular Formula:
C18H18F3NO2
InChi Key:
JDLSRXWHEBFHNC-UHFFFAOYSA-N
SMILES:
CCCCOC(=O)C1=CC=CC=C1NC1=CC=CC(=C1)C(F)(F)F
1
–
1
of
1
results
1
Butyl 2-[[3-(Trifluoromethyl)phenyl]-amino]benzoate (Butyl Flufenamate), Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.