accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COONH2NH2HClHCl

CAS RN 67396-08-1

IUPAC Name: methyl 2,6-diaminohexanoate dihydrochloride
Synonyms: methyl 2,6-diaminohexanoate dihydrochloride
Molecular Weight (g/mol): 233.13
Molecular Formula: C7H18Cl2N2O2
InChi Key: SXZCBVCQHOJXDR-UHFFFAOYNA-N
SMILES: Cl.Cl.COC(=O)C(N)CCCCN
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results