accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 674-26-0

IUPAC Name: 4-hydroxy-4-methyloxan-2-one
Synonyms: mevalonolactone
Molecular Weight (g/mol): 130.14
Molecular Formula: C6H10O3
InChi Key: JYVXNLLUYHCIIH-UHFFFAOYNA-N
SMILES: CC1(O)CCOC(=O)C1
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (6)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)