accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3CH3HOCH3CH3HOCH3CH3HODy

CAS RN 6742-68-3

IUPAC Name: tris(propan-2-ol) dysprosium
Synonyms: tris(isopropyl alcohol) dysprosium
Molecular Weight (g/mol): 342.79
Molecular Formula: C9H24DyO3
InChi Key: IWDABVYEISORRX-UHFFFAOYSA-N
SMILES: [Dy].CC(C)O.CC(C)O.CC(C)O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)