accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3CH3HOCH3CH3HOCH3CH3HOYb

CAS RN 6742-69-4

IUPAC Name: tris(propan-2-ol) ytterbium
Synonyms: tris(isopropyl alcohol) ytterbium
Molecular Weight (g/mol): 353.34
Molecular Formula: C9H24O3Yb
InChi Key: IVGMSSQGEVYWID-UHFFFAOYSA-N
SMILES: [Yb].CC(C)O.CC(C)O.CC(C)O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)