CAS RN 674289-55-5
IUPAC Name:
3a-hydroxy-6-methyl-1-phenyl-1H,2H,3H,3aH,4H-pyrrolo[2,3-b]quinolin-4-one
Synonyms:
3a-hydroxy-6-methyl-1-phenyl-2H,3H-pyrrolo[2,3-b]quinolin-4-one
Molecular Weight (g/mol):
292.34
Molecular Formula:
C18H16N2O2
InChi Key:
LZAXPYOBKSJSEX-UHFFFAOYNA-N
SMILES:
CC1=CC2=C(C=C1)N=C1N(CCC1(O)C2=O)C1=CC=CC=C1
1
–
2
of
2
results
1
(+/-)-Blebbistatin, TRC
CAS: 674289-55-5 Molecular Formula: C18 H16 N2 O2 Molecular Weight (g/mol): 292.33 IUPAC Name: 3a-hydroxy-6-methyl-1-phenyl-2,3-dihydropyrrolo[2,3-b]quinolin-4-one SMILES: Cc1ccc2N=C3N(CCC3(O)C(=O)c2c1)c4ccccc4
2
Promotions Available
Tocris Bioscience™ (±)-Blebbistatin
Tocris Bioscience™ (±)-Blebbistatin