accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 674792-05-3

CH3H3C(S)NHNOOH3CCH3H3C

CAS RN 674792-05-3

IUPAC Name: tert-butyl (2S)-2-(propan-2-yl)piperazine-1-carboxylate
Synonyms: tert-butyl (2S)-2-isopropylpiperazine-1-carboxylate
Molecular Weight (g/mol): 228.34
Molecular Formula: C12H24N2O2
InChi Key: NZTWGWFHWJARJX-SNVBAGLBSA-N
SMILES: CC(C)[C@H]1CNCCN1C(=O)OC(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results