accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HOBOHClCl

CAS RN 67492-50-6

IUPAC Name: (3,5-dichlorophenyl)boronic acid
Synonyms: 3,5-dichlorophenylboronic acid
Molecular Weight (g/mol): 190.81
Molecular Formula: C6H5BCl2O2
InChi Key: DKYRKAIKWFHQHM-UHFFFAOYSA-N
SMILES: OB(O)C1=CC(Cl)=CC(Cl)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results