accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 675-09-2

CH3OOCH3

CAS RN 675-09-2

IUPAC Name: 4,6-dimethyl-2H-pyran-2-one
Synonyms: 4,6-dimethyl-pyran-2-one
Molecular Weight (g/mol): 124.14
Molecular Formula: C7H8O2
InChi Key: IXYLIUKQQQXXON-UHFFFAOYSA-N
SMILES: CC1=CC(C)=CC(=O)O1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results