accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 677327-29-6

IUPAC Name: N-(5-iodo-3-methylpyridin-2-yl)-2,2-dimethylpropanamide
Synonyms: N-(5-iodo-3-methylpyridin-2-yl)-2,2-dimethylpropanamide
Molecular Weight (g/mol): 318.16
Molecular Formula: C11H15IN2O
InChi Key: LRQWADXLVLGBSY-UHFFFAOYSA-N
SMILES: CC1=CC(I)=CN=C1NC(=O)C(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results