accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OOClNCl

CAS RN 67754-03-4

IUPAC Name: methyl 2,5-dichloropyridine-3-carboxylate
Synonyms: methyl 2,5-dichloropyridine-3-carboxylate
Molecular Weight (g/mol): 206.02
Molecular Formula: C7H5Cl2NO2
InChi Key: CNPFXYWTMBTSSI-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(Cl)=CN=C1Cl
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (7)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)