accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3BrOCH3

CAS RN 67868-73-9

IUPAC Name: 4-bromo-2-methoxy-1-methylbenzene
Synonyms: 4-bromo-2-methoxy-1-methylbenzene
Molecular Weight (g/mol): 201.06
Molecular Formula: C8H9BrO
InChi Key: RAQYGVDRDNKTOM-UHFFFAOYSA-N
SMILES: COC1=C(C)C=CC(Br)=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results