CAS RN 67879-58-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Caftaric Acid, TRC
CAS: 67879-58-7 Molecular Formula: C13 H12 O9 Molecular Weight (g/mol): 312.23 Synonym: Butanedioic acid, 2-[[(2E)-3-(3,4-dihydroxyphenyl)-1-oxo-2-propenyl]oxy]-3-hydroxy-, (2R,3R)- (9CI),Butanedioic acid, 2-[[3-(3,4-dihydroxyphenyl)-1-oxo-2-propenyl]oxy]-3-hydroxy-, [R-[R*,R*-(E)]]-,(2R,3R)-2-[[(2E)-3-(3,4-Dihydroxyphenyl)-1-oxo-2-propen-1-yl]oxy]-3-hydroxybutanedioic acid,trans-Caftaric acid IUPAC Name: (2R,3R)-2-[(E)-3-(3,4-dihydroxyphenyl)prop-2-enoyl]oxy-3-hydroxybutanedioic acid SMILES: O[C@H]([C@@H](OC(=O)\C=C\c1ccc(O)c(O)c1)C(=O)O)C(=O)O