accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 67914-60-7

IUPAC Name: 1-[4-(4-hydroxyphenyl)piperazin-1-yl]ethan-1-one
Synonyms: 1-[4-(4-hydroxyphenyl)piperazin-1-yl]ethanone
Molecular Weight (g/mol): 220.27
Molecular Formula: C12H16N2O2
InChi Key: AGVNLFCRZULMKK-UHFFFAOYSA-N
SMILES: CC(=O)N1CCN(CC1)C1=CC=C(O)C=C1
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (2)
  • (2)
  • (2)
Industry Type 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)