accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BrO

CAS RN 6793-92-6

IUPAC Name: 1-(benzyloxy)-4-bromobenzene
Synonyms: 1-(benzyloxy)-4-bromobenzene
Molecular Weight (g/mol): 263.13
Molecular Formula: C13H11BrO
InChi Key: OUQSGILAXUXMGI-UHFFFAOYSA-N
SMILES: BrC1=CC=C(OCC2=CC=CC=C2)C=C1
Molecular Weight (g/mol) 
  • (3)
  • (3)
  • (2)
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)