accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NH2Cl

CAS RN 67952-93-6

IUPAC Name: 1-(3-chloro-4-methylphenyl)methanamine
Synonyms: 1-(3-chloro-4-methylphenyl)methanamine
Molecular Weight (g/mol): 155.63
Molecular Formula: C8H10ClN
InChi Key: MXIGALIASISPNU-UHFFFAOYSA-N
SMILES: CC1=CC=C(CN)C=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results