accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 680-15-9

IUPAC Name: methyl 2,2-difluoro-2-(fluorosulfonyl)acetate
Synonyms: methyl 2,2-difluoro-2-(fluorosulfonyl)acetate
Molecular Weight (g/mol): 192.11
Molecular Formula: C3H3F3O4S
InChi Key: GQJCAQADCPTHKN-UHFFFAOYSA-N
SMILES: COC(=O)C(F)(F)S(F)(=O)=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results