accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 68165-06-0

CH3NO

CAS RN 68165-06-0

IUPAC Name: 1-methylpyrrolidin-3-one
Synonyms: 1-methylpyrrolidin-3-one
Molecular Weight (g/mol): 99.13
Molecular Formula: C5H9NO
InChi Key: SLPUTJFVMJPMKV-UHFFFAOYSA-N
SMILES: CN1CCC(=O)C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results