CAS RN 6818-99-1
IUPAC Name:
5-chloro-1H-1,2,4-triazole
Synonyms:
3-chloro-2H-1,2,4-triazole
Molecular Weight (g/mol):
103.51
Molecular Formula:
C2H2ClN3
InChi Key:
QGOUKZPSCTVYLX-UHFFFAOYSA-N
SMILES:
ClC1=NC=NN1
1
–
1
of
1
results
1
3-Chloro-1,2,4-triazole, 97+%, Thermo Scientific™
CAS: 6818-99-1 Molecular Formula: C2H2ClN3 Molecular Weight (g/mol): 103.51 MDL Number: MFCD00128812 InChI Key: QGOUKZPSCTVYLX-UHFFFAOYSA-N Synonym: 3-chloro-1,2,4-triazole,3-chloro-1h-1,2,4-triazole,3-chloro-4h-1,2,4-triazole,1h-1,2,4-triazole, 3-chloro,3-chloro-s-triazole,s-triazole, 3-chloro,1h-1,2,4-triazole, 5-chloro,3-chloro-1h-1,2,4 triazole,3-chloro-2h-1,2,4-triazole PubChem CID: 81265 IUPAC Name: 5-chloro-1H-1,2,4-triazole SMILES: ClC1=NC=NN1