accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 683-72-7

H2NOClCl

CAS RN 683-72-7

IUPAC Name: 2,2-dichloroacetamide
Synonyms: dichloroacetamide
Molecular Weight (g/mol): 127.95
Molecular Formula: C2H3Cl2NO
InChi Key: WCGGWVOVFQNRRS-UHFFFAOYSA-N
SMILES: NC(=O)C(Cl)Cl
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results