accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 68302-57-8

IUPAC Name: 2-amino-5-oxo-7-(propan-2-yl)-5H-chromeno[2,3-b]pyridine-3-carboxylic acid
Synonyms: aphthasol amlenanox amlexanox (product) OraDisc solfa amlexanox (substance) amlexanox elics amlexanox [usan:inn:jan] amlexanoxum
Molecular Weight (g/mol): 298.3
Molecular Formula: C16H14N2O4
InChi Key: SGRYPYWGNKJSDL-UHFFFAOYSA-N
SMILES: CC(C)C1=CC=C2OC3=NC(N)=C(C=C3C(=O)C2=C1)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results