accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

FBrFFF

CAS RN 68322-84-9

IUPAC Name: 2-bromo-1-fluoro-4-(trifluoromethyl)benzene
Synonyms: 2-bromo-1-fluoro-4-(trifluoromethyl)benzene
Molecular Weight (g/mol): 243
Molecular Formula: C7H3BrF4
InChi Key: RZJOIMPUMMQKFR-UHFFFAOYSA-N
SMILES: FC1=CC=C(C=C1Br)C(F)(F)F
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)